2-Chloro-N-{1,4,6-trimethyl-1H-pyrazolo[3,4-b]pyridin-3-yl}acetamide, tech

Lieferant: Apollo Scientific

OR33155-1G
APOSOR33155-1GEA 1570 EUR
APOSOR33155-1G
2-Chloro-N-{1,4,6-trimethyl-1H-pyrazolo[3,4-b]pyridin-3-yl}acetamide, tech
2-Chloro-N-{1,4,6-trimethyl-1H-pyrazolo[3,4-b]pyridin-3-yl}acetamide
Formel: C₁₁H₁₃ClN₄O
Molecular Weight: 252,7 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03305741
CAS-Nummer: 866131-79-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR