N-(4-Acetylphenyl)-1,2,3,4-tetrahydroisoquinoline-2-carboxamide, tech

Lieferant: Apollo Scientific

OR33157-1G
APOSOR33157-1GEA 1570 EUR
APOSOR33157-1G
N-(4-Acetylphenyl)-1,2,3,4-tetrahydroisoquinoline-2-carboxamide, tech
N-(4-Acetylphenyl)-1,2,3,4-tetrahydroisoquinoline-2-carboxamide
Formel: C₁₈H₁₈N₂O₂
Molecular Weight: 294,348 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03102637
CAS-Nummer: 692732-82-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR