1-(4-Methoxy-2-nitrophenyl)-1H-pyrrole 95%
Lieferant: Apollo Scientific
OR33160-1G
APOSOR33160-1GEA
427
EUR
APOSOR33160-1G
1-(4-Methoxy-2-nitrophenyl)-1H-pyrrole 95%
1-(4-Methoxy-2-nitrophenyl)-1H-pyrrole
Formel:
C₁₁H₁₀N₂O₃ Molecular Weight: 218,209 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD03305765 CAS-Nummer: 59194-25-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...