3-(Pyrrolidine-1-carbonyl)-1H-indole 95%

Lieferant: Apollo Scientific

OR33163-1G
APOSOR33163-1GEA 1330 EUR
APOSOR33163-1G
3-(Pyrrolidine-1-carbonyl)-1H-indole 95%
3-(Pyrrolidine-1-carbonyl)-1H-indole
Formel: C₁₃H₁₄N₂O
Molecular Weight: 214,263 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03305785
CAS-Nummer: 27393-78-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR