Methyl-5-chloro-2-oxo-1,2-dihydro-3-pyridinecarboxylate 95%

Lieferant: Apollo Scientific

OR33168-1G
APOSOR33168-1GEA 188 EUR
APOSOR33168-1G
Methyl-5-chloro-2-oxo-1,2-dihydro-3-pyridinecarboxylate 95%
Methyl-5-chloro-2-oxo-1,2-dihydro-3-pyridinecarboxylate
Formel: C₇H₆ClNO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD04113615
CAS-Nummer: 886360-77-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR