9,9-Dimethyl-7,8,9,10-tetrahydrophenanthridin-7-one, tech

Lieferant: Apollo Scientific

OR33170-1G
APOSOR33170-1GEA 1570 EUR
APOSOR33170-1G
9,9-Dimethyl-7,8,9,10-tetrahydrophenanthridin-7-one, tech
9,9-Dimethyl-7,8,9,10-tetrahydrophenanthridin-7-one
Formel: C₁₅H₁₅NO
Molecular Weight: 225,286 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03848688
CAS-Nummer: 70062-80-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR