2,2,4-Trimethyl-2,3-dihydro-1H-1,5-benzodiazepine, tech

Lieferant: Apollo Scientific

OR33171-1G
APOSOR33171-1GEA 1570 EUR
APOSOR33171-1G
2,2,4-Trimethyl-2,3-dihydro-1H-1,5-benzodiazepine, tech
2,2,4-Trimethyl-2,3-dihydro-1H-1,5-benzodiazepine
Formel: C₁₂H₁₆N₂
Molecular Weight: 188,269 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00086184
CAS-Nummer: 24107-34-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR