2-(2,5-Dimethyl-1H-pyrrol-1-yl)-1,3-thiazole 95%

Lieferant: Apollo Scientific

OR33174-1G
APOSOR33174-1GEA 1120 EUR
APOSOR33174-1G
2-(2,5-Dimethyl-1H-pyrrol-1-yl)-1,3-thiazole 95%
2-(2,5-Dimethyl-1H-pyrrol-1-yl)-1,3-thiazole
Formel: C₉H₁₀N₂S
Molecular Weight: 178,254 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04126046
CAS-Nummer: 28142-85-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR