(2Z)-2-[(2,5-Dimethoxyphenyl)methylidene]-1-azabicyclo[2.2.2]octan-3-one, tech

Lieferant: Apollo Scientific

OR33176-1G
APOSOR33176-1GEA 1320 EUR
APOSOR33176-1G
(2Z)-2-[(2,5-Dimethoxyphenyl)methylidene]-1-azabicyclo[2.2.2]octan-3-one, tech
(2Z)-2-[(2,5-Dimethoxyphenyl)methylidene]-1-azabicyclo[2.2.2]octan-3-one
Formel: C₁₆H₁₉NO₃
Molecular Weight: 273,327 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02728993
CAS-Nummer: 315244-82-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR