3-[(4-Bromophenyl)methoxy]pyridin-2-amine, tech

Lieferant: Apollo Scientific

OR33180-1G
APOSOR33180-1GEA 1570 EUR
APOSOR33180-1G
3-[(4-Bromophenyl)methoxy]pyridin-2-amine, tech
3-[(4-Bromophenyl)methoxy]pyridin-2-amine
Formel: C₁₂H₁₁BrN₂O
Molecular Weight: 279,133 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD05256289
CAS-Nummer: 866135-55-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR