2-{6-Chloroimidazo[1,2-a]pyridin-2-yl}benzoic acid, tech

Lieferant: Apollo Scientific

OR33182-1G
APOSOR33182-1GEA 1570 EUR
APOSOR33182-1G
2-{6-Chloroimidazo[1,2-a]pyridin-2-yl}benzoic acid, tech
2-{6-Chloroimidazo[1,2-a]pyridin-2-yl}benzoic acid
Formel: C₁₄H₉ClN₂O₂
Molecular Weight: 272,686 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06496192
CAS-Nummer: 900015-65-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR