(2E)-1-{6-Chloro-2-methylimidazo[1,2-a]pyridin-3-yl}-3-(dimethylamino)prop-2-en-1-one, tech

Lieferant: Apollo Scientific

OR33183-1G
APOSOR33183-1GEA 1570 EUR
APOSOR33183-1G
(2E)-1-{6-Chloro-2-methylimidazo[1,2-a]pyridin-3-yl}-3-(dimethylamino)prop-2-en-1-one, tech
(2E)-1-{6-Chloro-2-methylimidazo[1,2-a]pyridin-3-yl}-3-(dimethylamino)prop-2-en-1-one
Formel: C₁₃H₁₄ClN₃O
Molecular Weight: 263,723 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06496193
CAS-Nummer: 900015-66-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR