3-Bromo-8-methylimidazo[1,2-a]pyridine 95%

Lieferant: Apollo Scientific
Achtung

OR33184-1G
APOSOR33184-1GEA 190 EUR
APOSOR33184-1G
3-Bromo-8-methylimidazo[1,2-a]pyridine 95%
3-Bromo-8-methylimidazo[1,2-a]pyridine
Formel: C₈H₇BrN₂
Molecular Weight: 211,059 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06496194
CAS-Nummer: 866135-66-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR