1-(3,4-Dimethoxyphenyl)-1H-pyrrole-2-carbaldehyde, tech
Lieferant: Apollo Scientific
OR33186-1G
APOSOR33186-1GEA
1570
EUR
APOSOR33186-1G
1-(3,4-Dimethoxyphenyl)-1H-pyrrole-2-carbaldehyde, tech
1-(3,4-Dimethoxyphenyl)-1H-pyrrole-2-carbaldehyde
Formel:
C₁₃H₁₃NO₃ Molecular Weight: 231,247 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD05256301 CAS-Nummer: 43053-78-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...