3-Bromo-6-methylimidazo[1,2-a]pyridine 95%

Lieferant: Apollo Scientific

OR33187-1G
APOSOR33187-1GEA 671 EUR
APOSOR33187-1G
3-Bromo-6-methylimidazo[1,2-a]pyridine 95%
3-Bromo-6-methylimidazo[1,2-a]pyridine
Formel: C₈H₇BrN₂
Lagertemperatur: Raumtemperatur
MDL: MFCD06496195
CAS-Nummer: 866135-71-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR