6-Bromo-8-methylimidazo[1,2-a]pyridine-2-carboxylic acid 95%
Lieferant: Apollo Scientific
OR33189-1G
APOSOR33189-1GEA
427
EUR
APOSOR33189-1G
6-Bromo-8-methylimidazo[1,2-a]pyridine-2-carboxylic acid 95%
6-Bromo-8-methylimidazo[1,2-a]pyridine-2-carboxylic acid
Formel:
C₉H₇BrN₂O₂ Molecular Weight: 255,068 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD06496201 CAS-Nummer: 866135-83-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...