Ethyl 2-[(4-methylphenyl)sulphanyl]acetate 95%

Lieferant: Apollo Scientific

OR33191-1G
APOSOR33191-1GEA 224 EUR
APOSOR33191-1G
Ethyl 2-[(4-methylphenyl)sulphanyl]acetate 95%
Ethyl 2-[(4-methylphenyl)sulphanyl]acetate
Formel: C₁₁H₁₄O₂S
Molecular Weight: 210.293 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04275108
CAS-Nummer: 14738-27-3

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR