Methyl-4-bromo-5-methyl-1H-pyrazole-3-carboxylate 95%

Lieferant: Apollo Scientific

OR33202-1G
APOSOR33202-1GEA 336 EUR
APOSOR33202-1G
Methyl-4-bromo-5-methyl-1H-pyrazole-3-carboxylate 95%
Methyl-4-bromo-5-methyl-1H-pyrazole-3-carboxylate
Formel: C₆H₇BrN₂O₂
Molecular Weight: 219.04 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08056667
CAS-Nummer: 1232838-31-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR