2-Chloro-4-(3-methyl-1H-pyrazol-1-yl)benzenecarboxylic acid 95%

Lieferant: Apollo Scientific

Synonyme: 2-Chloro-4-(3-methyl-1H-pyrazol-1-yl)benzoic acid

OR33205-1G
APOSOR33205-1GEA 671 EUR
APOSOR33205-1G
2-Chloro-4-(3-methyl-1H-pyrazol-1-yl)benzenecarboxylic acid 95%
2-Chloro-4-(3-methyl-1H-pyrazol-1-yl)benzenecarboxylic acid
Formel: C₁₁H₉ClN₂O₂
Molecular Weight: 236,66 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08056672
CAS-Nummer: 220461-68-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR