2,6-Bis(1H-imidazol-1-yl)benzonitrile, tech

Lieferant: Apollo Scientific

OR33206-1G
APOSOR33206-1GEA 1570 EUR
APOSOR33206-1G
2,6-Bis(1H-imidazol-1-yl)benzonitrile, tech
2,6-Bis(1H-imidazol-1-yl)benzonitrile
Formel: C₁₃H₉N₅
Molecular Weight: 235,244 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00172157
CAS-Nummer: 285987-29-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR