1-[2-(4-Chlorophenyl)-2-oxoethyl]-1,2-dihydropyridin-2-one, tech

Lieferant: Apollo Scientific

OR33208-1G
APOSOR33208-1GEA 1570 EUR
APOSOR33208-1G
1-[2-(4-Chlorophenyl)-2-oxoethyl]-1,2-dihydropyridin-2-one, tech
1-[2-(4-Chlorophenyl)-2-oxoethyl]-1,2-dihydropyridin-2-one
Formel: C₁₃H₁₀ClNO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00139074
CAS-Nummer: 108664-55-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR