3-(4-Chlorophenoxy)butan-2-one, tech
Lieferant: Apollo Scientific
OR33209-1G
APOSOR33209-1GEA
1570
EUR
APOSOR33209-1G
3-(4-Chlorophenoxy)butan-2-one, tech
3-(4-Chlorophenoxy)butan-2-one
Formel:
C₁₀H₁₁ClO₂ Molecular Weight: 198,646 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00243668 CAS-Nummer: 3782-11-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...