6-[(2-Methylphenyl)methyl]pyridazin-3-ol, tech

Lieferant: Apollo Scientific

OR33212-1G
APOSOR33212-1GEA 1570 EUR
APOSOR33212-1G
6-[(2-Methylphenyl)methyl]pyridazin-3-ol, tech
6-[(2-Methylphenyl)methyl]pyridazin-3-ol
Formel: C₁₂H₁₂N₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD00214802
CAS-Nummer: 60932-69-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR