N-(4-Methoxyphenyl)-2-oxo-1-(prop-2-yn-1-yl)-1,2-dihydropyridine-3-carboxamide, tech

Lieferant: Apollo Scientific

OR33219-1G
APOSOR33219-1GEA 1570 EUR
APOSOR33219-1G
N-(4-Methoxyphenyl)-2-oxo-1-(prop-2-yn-1-yl)-1,2-dihydropyridine-3-carboxamide, tech
N-(4-Methoxyphenyl)-2-oxo-1-(prop-2-yn-1-yl)-1,2-dihydropyridine-3-carboxamide
Formel: C₁₆H₁₄N₂O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD00141338
CAS-Nummer: 339028-30-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR