5-Chloro-2-oxo-N-phenyl-1-(prop-2-yn-1-yl)-1,2-dihydropyridine-3-carboxamide, tech

Lieferant: Apollo Scientific

OR33220-1G
APOSOR33220-1GEA 1570 EUR
APOSOR33220-1G
5-Chloro-2-oxo-N-phenyl-1-(prop-2-yn-1-yl)-1,2-dihydropyridine-3-carboxamide, tech
5-Chloro-2-oxo-N-phenyl-1-(prop-2-yn-1-yl)-1,2-dihydropyridine-3-carboxamide
Formel: C₁₅H₁₁ClN₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00141340
CAS-Nummer: 339028-41-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR