2-(2-Chlorophenyl)-2-(morpholin-4-yl)acetonitrile 95%

Lieferant: Apollo Scientific

OR33226-1G
APOSOR33226-1GEA 671 EUR
APOSOR33226-1G
2-(2-Chlorophenyl)-2-(morpholin-4-yl)acetonitrile 95%
2-(2-Chlorophenyl)-2-(morpholin-4-yl)acetonitrile
Formel: C₁₂H₁₃ClN₂O
Molecular Weight: 236,697 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00173632
CAS-Nummer: 66549-12-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR