1,3-Dimethyl-5-(prop-2-en-1-yloxy)-1H-pyrazole-4-carbaldehyde 95%
Lieferant: Apollo Scientific
OR33228-1G
APOSOR33228-1GEA
793
EUR
APOSOR33228-1G
1,3-Dimethyl-5-(prop-2-en-1-yloxy)-1H-pyrazole-4-carbaldehyde 95%
1,3-Dimethyl-5-(prop-2-en-1-yloxy)-1H-pyrazole-4-carbaldehyde
Formel:
C₉H₁₂N₂O₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00665212 CAS-Nummer: 321574-23-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...