11,13-Dimethoxy-3-methyl-4,8-dioxa-3-azatricyclo[7.4.0.02,6]trideca-1(13),9,11-triene, tech
Lieferant: Apollo Scientific
OR33229-1G
APOSOR33229-1GEA
1570
EUR
APOSOR33229-1G
11,13-Dimethoxy-3-methyl-4,8-dioxa-3-azatricyclo[7.4.0.02,6]trideca-1(13),9,11-triene, tech
11,13-Dimethoxy-3-methyl-4,8-dioxa-3-azatricyclo[7.4.0.02,6]trideca-1(13),9,11-triene
Formel:
C₁₃H₁₇NO₄ Molecular Weight: 251,278 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00232822 CAS-Nummer: 400088-00-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...