(2R,6S)-12-Bromo-3-methyl-4,8-dioxa-3-azatricyclo[7.4.0.02,6]trideca-1(13),9,11-triene, tech

Lieferant: Apollo Scientific

OR33231-1G
APOSOR33231-1GEA 1570 EUR
APOSOR33231-1G
(2R,6S)-12-Bromo-3-methyl-4,8-dioxa-3-azatricyclo[7.4.0.02,6]trideca-1(13),9,11-triene, tech
(2R,6S)-12-Bromo-3-methyl-4,8-dioxa-3-azatricyclo[7.4.0.02,6]trideca-1(13),9,11-triene
Formel: C₁₁H₁₂BrNO₂
Molecular Weight: 270,122 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00231697
CAS-Nummer: 321574-26-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR