(3E)-3-{[(4-Chlorophenyl)methoxy]imino}propanenitrile, tech
Lieferant: Apollo Scientific
OR33232-1G
APOSOR33232-1GEA
1570
EUR
APOSOR33232-1G
(3E)-3-{[(4-Chlorophenyl)methoxy]imino}propanenitrile, tech
(3E)-3-{[(4-Chlorophenyl)methoxy]imino}propanenitrile
Formel:
C₁₀H₉ClN₂O Molecular Weight: 208,644 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00663745 CAS-Nummer: 339096-99-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...