(2Z)-4-Chloro-2-{[(4-chlorophenyl)amino]methylidene}-3-oxobutanenitrile, tech

Lieferant: Apollo Scientific

OR33233-1G
APOSOR33233-1GEA 1570 EUR
APOSOR33233-1G
(2Z)-4-Chloro-2-{[(4-chlorophenyl)amino]methylidene}-3-oxobutanenitrile, tech
(2Z)-4-Chloro-2-{[(4-chlorophenyl)amino]methylidene}-3-oxobutanenitrile
Formel: C₁₁H₈Cl₂N₂O
Molecular Weight: 255,1 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00663748
CAS-Nummer: 339097-10-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR