2-(2,4-Dichlorophenyl)-2-(morpholin-4-yl)acetonitrile, tech

Lieferant: Apollo Scientific

OR33234-1G
APOSOR33234-1GEA 1570 EUR
APOSOR33234-1G
2-(2,4-Dichlorophenyl)-2-(morpholin-4-yl)acetonitrile, tech
2-(2,4-Dichlorophenyl)-2-(morpholin-4-yl)acetonitrile
Formel: C₁₂H₁₂Cl₂N₂O
Molecular Weight: 271,142 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00172631
CAS-Nummer: 135595-04-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR