Methyl-4-[3-(4-piperidinyl)-1H-pyrazol-5-yl]phenyl ether 95%
Lieferant: Apollo Scientific
Synonyme:
4-[5-(4-Methoxyphenyl)-1H-pyrazol-3-yl]piperidine
OR33241-1G
APOSOR33241-1GEA
671
EUR
APOSOR33241-1G
Methyl-4-[3-(4-piperidinyl)-1H-pyrazol-5-yl]phenyl ether 95%
Methyl-4-[3-(4-piperidinyl)-1H-pyrazol-5-yl]phenyl ether
Formel:
C₁₅H₁₉N₃O Molecular Weight: 257,34 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00665098 CAS-Nummer: 103660-47-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...