Methyl 2-(3,3-dimethyl-2-oxoazetidin-1-yl)benzoate, tech

Lieferant: Apollo Scientific

OR33244-1G
APOSOR33244-1GEA 1570 EUR
APOSOR33244-1G
Methyl 2-(3,3-dimethyl-2-oxoazetidin-1-yl)benzoate, tech
Methyl 2-(3,3-dimethyl-2-oxoazetidin-1-yl)benzoate
Formel: C₁₃H₁₅NO₃
Molecular Weight: 233,263 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00793431
CAS-Nummer: 339100-88-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR