(2E)-2-[2-(2,3-Dihydro-1,4-benzodioxin-6-yl)hydrazin-1-ylidene]cyclohexan-1-one, tech

Lieferant: Apollo Scientific

OR33246-1G
APOSOR33246-1GEA 1570 EUR
APOSOR33246-1G
(2E)-2-[2-(2,3-Dihydro-1,4-benzodioxin-6-yl)hydrazin-1-ylidene]cyclohexan-1-one, tech
(2E)-2-[2-(2,3-Dihydro-1,4-benzodioxin-6-yl)hydrazin-1-ylidene]cyclohexan-1-one
Formel: C₁₄H₁₆N₂O₃
Molecular Weight: 260.288 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01568696
CAS-Nummer: 339101-62-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR