4-(Dimethylamino)-2-(phenylamino)pyridine-3-carbonitrile, tech

Lieferant: Apollo Scientific

OR33247-1G
APOSOR33247-1GEA 1570 EUR
APOSOR33247-1G
4-(Dimethylamino)-2-(phenylamino)pyridine-3-carbonitrile, tech
4-(Dimethylamino)-2-(phenylamino)pyridine-3-carbonitrile
Formel: C₁₄H₁₄N₄
Molecular Weight: 238,288 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01314736
CAS-Nummer: 339102-77-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR