Methyl 3-(4-chlorophenyl)-4,5-dihydro-1,2-oxazole-5-carboxylate 95%

Lieferant: Apollo Scientific

OR33248-1G
APOSOR33248-1GEA 427 EUR
APOSOR33248-1G
Methyl 3-(4-chlorophenyl)-4,5-dihydro-1,2-oxazole-5-carboxylate 95%
Methyl 3-(4-chlorophenyl)-4,5-dihydro-1,2-oxazole-5-carboxylate
Formel: C₁₁H₁₀ClNO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD02185946
CAS-Nummer: 91258-47-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR