2-(Methylamino)-4-phenylpyridine-3-carbonitrile, tech

Lieferant: Apollo Scientific

OR33249-1G
APOSOR33249-1GEA 1570 EUR
APOSOR33249-1G
2-(Methylamino)-4-phenylpyridine-3-carbonitrile, tech
2-(Methylamino)-4-phenylpyridine-3-carbonitrile
Formel: C₁₃H₁₁N₃
Molecular Weight: 209,247 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01444225
CAS-Nummer: 339103-54-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR