5H,6H,7H,8H-Imidazo[1,5-c]pyrimidin-5-one, tech

Lieferant: Apollo Scientific

Synonyme: 7,8-Dihydroimidazo[1,5-c]pyrimidin-5(6H)-one

OR33250-1G
APOSOR33250-1GEA 133 EUR
APOSOR33250-1G
5H,6H,7H,8H-Imidazo[1,5-c]pyrimidin-5-one, tech
5H,6H,7H,8H-Imidazo[1,5-c]pyrimidin-5-one
Formel: C₆H₇N₃O
Molecular Weight: 137,139 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01443896
CAS-Nummer: 14509-66-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR