(2E)-3-[5-(Hydroxymethyl)furan-2-yl]-1-phenylprop-2-en-1-one, tech

Lieferant: Apollo Scientific

OR33252-1G
APOSOR33252-1GEA 1570 EUR
APOSOR33252-1G
(2E)-3-[5-(Hydroxymethyl)furan-2-yl]-1-phenylprop-2-en-1-one, tech
(2E)-3-[5-(Hydroxymethyl)furan-2-yl]-1-phenylprop-2-en-1-one
Formel: C₁₄H₁₂O₃
Molecular Weight: 228,243 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01315260
CAS-Nummer: 132391-04-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR