(2E)-3-(Dimethylamino)-1-[3-(pyrimidin-2-yloxy)phenyl]prop-2-en-1-one, tech

Lieferant: Apollo Scientific

OR33255-1G
APOSOR33255-1GEA 1570 EUR
APOSOR33255-1G
(2E)-3-(Dimethylamino)-1-[3-(pyrimidin-2-yloxy)phenyl]prop-2-en-1-one, tech
(2E)-3-(Dimethylamino)-1-[3-(pyrimidin-2-yloxy)phenyl]prop-2-en-1-one
Formel: C₁₅H₁₅N₃O₂
Molecular Weight: 269,299 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01568702
CAS-Nummer: 339105-45-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR