(3Z)-3-[(Dimethylamino)methylidene]-6-methyl-3,4-dihydro-2H-1-benzothiopyran-4-one, tech

Lieferant: Apollo Scientific

OR33258-1G
APOSOR33258-1GEA 1570 EUR
APOSOR33258-1G
(3Z)-3-[(Dimethylamino)methylidene]-6-methyl-3,4-dihydro-2H-1-benzothiopyran-4-one, tech
(3Z)-3-[(Dimethylamino)methylidene]-6-methyl-3,4-dihydro-2H-1-benzothiopyran-4-one
Formel: C₁₃H₁₅NOS
Molecular Weight: 233,329 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04126087
CAS-Nummer: 866136-49-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR