1-[4-Methyl-2-(morpholin-4-yl)quinolin-3-yl]ethan-1-one, tech

Lieferant: Apollo Scientific

OR33259-1G
APOSOR33259-1GEA 1570 EUR
APOSOR33259-1G
1-[4-Methyl-2-(morpholin-4-yl)quinolin-3-yl]ethan-1-one, tech
1-[4-Methyl-2-(morpholin-4-yl)quinolin-3-yl]ethan-1-one
Formel: C₁₆H₁₈N₂O₂
Molecular Weight: 270,326 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01568465
CAS-Nummer: 339106-15-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR