4-(N'-Hydroxycarbamimidoyl)benzeneboronic acid 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: (4-(N'-Hydroxycarbamimidoyl)phenyl)boronic acid

OR3326-1G
APOSOR3326-1GEA 148 EUR
APOSOR3326-1G
4-(N'-Hydroxycarbamimidoyl)benzeneboronic acid 95%
4-(N'-Hydroxycarbamimidoyl)benzeneboronic acid
Formel: C₇H₉BN₂O₃
Molecular Weight: 179.969 g/mol
Schmelzpunkt: 290…292 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD08436061
CAS-Nummer: 913835-61-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR