4-[(E)-2-(Dimethylamino)ethenyl]-2-oxo-2H-chromene-3-carbonitrile, tech

Lieferant: Apollo Scientific

OR33260-1G
APOSOR33260-1GEA 1570 EUR
APOSOR33260-1G
4-[(E)-2-(Dimethylamino)ethenyl]-2-oxo-2H-chromene-3-carbonitrile, tech
4-[(E)-2-(Dimethylamino)ethenyl]-2-oxo-2H-chromene-3-carbonitrile
Formel: C₁₄H₁₂N₂O₂
Molecular Weight: 240,257 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01304441
CAS-Nummer: 209617-29-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR