3-Benzyl-4-hydroxy-1-(2-methoxyethyl)-1,2-dihydropyridin-2-one, tech

Lieferant: Apollo Scientific

OR33262-1G
APOSOR33262-1GEA 1570 EUR
APOSOR33262-1G
3-Benzyl-4-hydroxy-1-(2-methoxyethyl)-1,2-dihydropyridin-2-one, tech
3-Benzyl-4-hydroxy-1-(2-methoxyethyl)-1,2-dihydropyridin-2-one
Formel: C₁₅H₁₇NO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD01568818
CAS-Nummer: 339106-88-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR