7-Methoxy-1-benzofuran-2-carbonitrile, tech

Lieferant: Apollo Scientific

OR33265-1G
APOSOR33265-1GEA 1570 EUR
APOSOR33265-1G
7-Methoxy-1-benzofuran-2-carbonitrile, tech
7-Methoxy-1-benzofuran-2-carbonitrile
Formel: C₁₀H₇NO₂
Molecular Weight: 173,168 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02082422
CAS-Nummer: 35351-49-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR