3-Chloro-6-(thiophen-2-yl)pyridazine 95%

Lieferant: Apollo Scientific

OR33268-1G
APOSOR33268-1GEA 364 EUR
APOSOR33268-1G
3-Chloro-6-(thiophen-2-yl)pyridazine 95%
3-Chloro-6-(thiophen-2-yl)pyridazine
Formel: C₈H₅ClN₂S
Molecular Weight: 196,657 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00114988
CAS-Nummer: 28657-41-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR