2-Chlor-5-nitrophenylboronsäure
Lieferant: Apollo Scientific
|
Achtung
|
OR3327-1G
OR3327-5G
OR3327-25G
APOSOR3327-1GEA
21.9
EUR
APOSOR3327-1G
APOSOR3327-5G
APOSOR3327-25G
2-Chlor-5-nitrophenylboronsäure
2-Chlor-5-nitrophenylboronsäure
Formel:
C₆H₅BClNO₄ Schmelzpunkt: 236…238 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD03095323 CAS-Nummer: 867333-29-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...