Methyl 4-(3-methylbutanoyl)-1H-pyrrole-2-carboxylate, tech

Lieferant: Apollo Scientific

OR33277-1G
APOSOR33277-1GEA 1570 EUR
APOSOR33277-1G
Methyl 4-(3-methylbutanoyl)-1H-pyrrole-2-carboxylate, tech
Methyl 4-(3-methylbutanoyl)-1H-pyrrole-2-carboxylate
Formel: C₁₁H₁₅NO₃
Molecular Weight: 209,242 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02571059
CAS-Nummer: 478259-48-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR